missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Kv11.2 Recombinant Protein Antigen

Product Code. 18328885 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328885 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328885 Supplier Novus Biologicals™ Supplier No. NBP317875PEP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Antibody Competition
Formulation PBS and 1M Urea, pH 7.4.
Gene ID (Entrez) 81033
Molecular Weight (g/mol) M.W. Theoretical: 25 kDa
Quantity 100 μg
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Gene Alias eag related protein 2, eag-related gene member 2, Eag-related protein 2, erg2, ERG-2, ether-a-go-go related gene potassium channel 2, Ether-a-go-go-related gene potassium channel 2, Ether-a-go-go-related protein 2, hERG-2, HERG2, Kv11.2, potassium voltage-gated channel subfamily H member 6, potassium voltage-gated channel, subfamily H (eag-related), member 6, Voltage-gated potassium channel subunit Kv11.2
Gene Symbol KCNH6
Research Category Neuroscience
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein Kv11.2
Show More Show Less
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.