missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv11.1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-80513
This item is not returnable.
View return policy
Description
Kv11.1 Polyclonal specifically detects Kv11.1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Kv11.1 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:1000 - 1:2000, Immunohistochemistry-Paraffin 1:1000 - 1:2000 | |
| Eag homolog, Eag-related protein 1, ERG, erg1, ERG-1, Ether-a-go-go-related gene potassium channel 1, ether-a-go-go-related potassium channel protein, Ether-a-go-go-related protein 1, H-ERG, HERG1, HERGhERG-1, Kv11.1, LQT2, potassium voltage-gated channel subfamily H member 2, potassium voltage-gated channel, subfamily H (eag-related), member 2, SQT1, Voltage-gated potassium channel subunit Kv11.1 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Lyophilized from whole serum | |
| KCNH2 | |
| A synthetic peptide from amino acid region 1120-1162 of mouse Kv11.1 conjugated to blue carrier protein was used as the antigen. The peptide is homologous in rat and human. | |
| 0.1 mL | |
| Neuroscience, Neurotransmission, Potassium Channels | |
| 3757 | |
| Human, Mouse, Rat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction