missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Kv1.6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30823-100ug
This item is not returnable.
View return policy
Description
Kv1.6 Polyclonal antibody specifically detects Kv1.6 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| Kv1.6 | |
| Polyclonal | |
| Western Blot : 5-10 μg/mL | |
| FLJ25134, HBK2, human brain potassium channel-2, KV1.6, potassium voltage-gated channel subfamily A member 6, potassium voltage-gated channel, shaker-related subfamily, member 6, Voltage-gated potassium channel HBK2, voltage-gated potassium channel protein Kv1.6, Voltage-gated potassium channel subunit Kv1.6 | |
| Synthetic peptide derived from the rat Kv1.6 conjugated to KLH | |
| 100 μg | |
| Signal Transduction | |
| 3742 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction