missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv1.6 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-98145CL1
This item is not returnable.
View return policy
Description
Kv1.6 Polyclonal antibody specifically detects Kv1.6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Kv1.6 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| FLJ25134, HBK2, human brain potassium channel-2, KV1.6, potassium voltage-gated channel subfamily A member 6, potassium voltage-gated channel, shaker-related subfamily, member 6, Voltage-gated potassium channel HBK2, voltage-gated potassium channel protein Kv1.6, Voltage-gated potassium channel subunit Kv1.6 | |
| Produced in rabbits immunized with E. coli-derived Human Kv1.6 fragment. | |
| 0.1 mL | |
| Signal Transduction | |
| 3742 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction