missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KV1.5 (KCNA5) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101762
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KCNA5.
In neurons voltage-dependent potassium channels are key determinants of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.
Specifications
| KV1.5 (KCNA5) | |
| Polyclonal | |
| Unconjugated | |
| Kcna5 | |
| ATFB7; cardiac potassium channel; delayed rectifier K+ channel; HCK1; HK2; HPCN1; insulinoma and islet potassium channel; KCNA5; Kv1; KV1.5; KV1-5; MGC117058; MGC117059; PCN1; Potassium (K+) channel protein alpha 5; potassium channel 1; potassium channel, voltage gated shaker related subfamily A, member 5; potassium voltage gated channel, shaker related subfamily, member 5; potassium voltage-gated channel subfamily A member 5; potassium voltage-gated channel, shaker-related subfamily, member 5; RCK7; RK4; voltage-gated potassium channel HK2; voltage-gated potassium channel protein Kv1.5; voltage-gated potassium channel subunit Kv1.5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16493, 25470, 3741 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P19024, P22460, Q61762 | |
| Kcna5 | |
| A synthesized peptide derived from human KCNA5(Accession P22460), corresponding to amino acid residues I252-G302. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction