missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KV1.4 (KCNA4) Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585937
This item is not returnable.
View return policy
Description
KV1.4 (KCNA4) Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, Flow
Potassium voltage-gated channel subfamily A member 4, also known as Kv1.4 or PCN2, is a protein that in humans is encoded by the KCNA4 gene. This gene encodes a member of the potassium channel, voltage-gated, shaker-related subfamily. It is mapped to 11p14.1. KCNA4 belongs to the A-type potassium current class, the members of which may be important in the regulation of the fast repolarizing phase of action potentials in heart and thus may influence the duration of cardiac action potential. KCNA4 also contributes to the cardiac transient outward potassium current (Ito1), the main contributing current to the repolarizing phase 1 of the cardiac action potential. This gene has been shown to interact with DLG4, KCNA2 and DLG1.
Specifications
| KV1.4 (KCNA4) | |
| Polyclonal | |
| Unconjugated | |
| KCNA4 | |
| cardiac potassium channel; fetal skeletal muscle potassium channel; HBK4; HK1; HPCN2; HUKII; KCHAN; Kcna4; KCNA4L; KCNA8; Kv1.4; Kv4; PCN2; potassium channel 2; potassium channel, voltage gated shaker related subfamily A, member 4; potassium voltage gated channel, shaker related subfamily, member 4; potassium voltage-gated channel subfamily A member 4; potassium voltage-gated channel, shaker-related subfamily, member 4; rapidly inactivating potassium channel; RCK4; RHK1; RK3; shaker-related potassium channel Kv1.4; type A potassium channel; voltage-gated K(+) channel HuKII; Voltage-gated potassium channel HBK4; voltage-gated potassium channel HK1; Voltage-gated potassium channel subunit Kv1.4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16492, 25469, 3739 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P15385, P22459, Q61423 | |
| KCNA4 | |
| Synthetic peptide within mouse Kv14 aa 201-250. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction