missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv1.3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£185.00 - £381.00
Specifications
| Antigen | Kv1.3 |
|---|---|
| Dilution | Western Blot 5 - 10 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18315232
|
Bio-Techne
NBP3-11898-25UG |
25 μg |
£185.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18332303
|
Novus Biologicals
NBP3-11898-100UG |
100 μg |
£381.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kv1.3 Polyclonal antibody specifically detects Kv1.3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Kv1.3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| HGK5, HLK3KV1.3, HPCN3PCN3, HUKIII, Kv1.3, MK3, potassium channel 3, potassium voltage-gated channel subfamily A member 3, potassium voltage-gated channel, shaker-related subfamily, member 3, type n potassium channel, Voltage-gated K(+) channel HuKIII, voltage-gated potassium channel protein Kv1.3, Voltage-gated potassium channel subunit Kv1.3 | |
| Synthetic peptide derived from the rat Kv1.3 potassium channel conjugated to KLH | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot 5 - 10 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS | |
| 3738 | |
| IgG | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title