missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Kv1.3 Polyclonal antibody specifically detects Kv1.3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Kv1.3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 5 - 10 ug/ml |
| Formulation | PBS |
| Gene Alias | HGK5, HLK3KV1.3, HPCN3PCN3, HUKIII, Kv1.3, MK3, potassium channel 3, potassium voltage-gated channel subfamily A member 3, potassium voltage-gated channel, shaker-related subfamily, member 3, type n potassium channel, Voltage-gated K(+) channel HuKIII, voltage-gated potassium channel protein Kv1.3, Voltage-gated potassium channel subunit Kv1.3 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide derived from the rat Kv1.3 potassium channel conjugated to KLH |
| Purification Method | Ammonium sulfate precipitation |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?