missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KV1.3 (KCNA3) Polyclonal Antibody
GREENER_CHOICE

Product Code. 17288263 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17288263 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17288263 Supplier Invitrogen™ Supplier No. PA5115486

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Kv1.3/KCNA3.

In neurons voltage-dependent potassium channels are key determinants of the resting membrane potential and of membrane excitability, conditioning the frequency, and shape of action potential.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KV1.3 (KCNA3)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene KCNA3
Gene Accession No. P16390, P22001
Gene Alias HGK5; HLK3; HPCN3; HUK3; HUKIII; Kca1-3; Kcna3; Kv1.3; KV3; MK3; Mk-3; PCN3; potassium channel 3; potassium channel, voltage gated shaker related subfamily A, member 3; potassium voltage gated channel, shaker related subfamily, member 3; potassium voltage-gated channel subfamily A member 3; potassium voltage-gated channel, shaker-related subfamily, member 3; RCK3; RGK5; type n potassium channel; voltage-gated K(+) channel HuKIII; voltage-gated potassium channel protein Kv1.3; Voltage-gated potassium channel protein Kv1.3 (RGK5) (RCK3) (KV3); voltage-gated potassium channel subunit Kv1.3; Voltage-gated potassium channel subunit Kv3
Gene Symbols KCNA3
Host Species Rabbit
Immunogen A synthesized peptide derived from human KCNA3(Accession P22001), corresponding to amino acid residues I244-P294.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16491, 3738
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.