missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kv1.3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48533
This item is not returnable.
View return policy
Description
Kv1.3 Polyclonal antibody specifically detects Kv1.3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Kv1.3 | |
| Polyclonal | |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| HGK5, HLK3KV1.3, HPCN3PCN3, HUKIII, Kv1.3, MK3, potassium channel 3, potassium voltage-gated channel subfamily A member 3, potassium voltage-gated channel, shaker-related subfamily, member 3, type n potassium channel, Voltage-gated K(+) channel HuKIII, voltage-gated potassium channel protein Kv1.3, Voltage-gated potassium channel subunit Kv1.3 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: EEQSQYMHVGSCQHLSSSAEELRKARSNSTLSKSEYMVIEEGGMNHSAFPQTPFKTGNSTATCTTNNNPNSCVNIK | |
| 0.1 mL | |
| Signal Transduction | |
| 3738 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction