Learn More
KV1.2 (KCNA2) Recombinant Rabbit Monoclonal Antibody (HL3163)
GREENER_CHOICE
KV1.2 (KCNA2) Recombinant Rabbit Monoclonal Antibody (HL3163)
Brand:  Invitrogen MA556774
Product Code. 30275822
  • £434.00 / 100µL
Description
KV1.2 (KCNA2) Recombinant Monoclonal Antibody for WB, IHC (P)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KV1.2 (KCNA2)
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P63141, P63142
KCNA2
The immunogen used to generate this antibody corresponds to human Kv1.2.
100 μL
Primary
Mouse, Rat
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
HL3163
Unconjugated
KCNA2
Akr6a4; BK2; CSMK1; delayed rectifier K+ channel; EIEE32; ENSMUSG00000074335; Gm10672; HBK5; HK4; HUKIV; k(v)1.2; KC22; Kca1-2; kcna2; kcna2.S; kcna2-a; KV1.2; Kv1.2' potassium channel; LOC100537815; MK2; Mk-2; NGK1; Potassium (K+) channel protein alpha 2, voltage dependent; potassium channel (BGK5); potassium channel subunit Kv 1.2; potassium channel, voltage gated shaker related subfamily A, member 2; potassium channel, voltage gated shaker related subfamily A, member 2 S homeolog; potassium voltage gated channel shaker related subfamily member 2; potassium voltage-gated channel subfamily A member 2; potassium voltage-gated channel, shaker-related subfamily, member 2; potassium voltage-gated channel, shaker-related subfamily, member 2 a; RAK; RBK2; RCK5; RP11-284N8.1; voltage-dependent K channel; voltage-gated channel, shaker-related subfamily, member 2; voltage-gated K(+) channel HuKIV; Voltage-gated potassium channel HBK5; voltage-gated potassium channel isoform 2; voltage-gated potassium channel isoform 3; voltage-gated potassium channel protein Kv1.2; Voltage-gated potassium channel subunit Kv1.2; XELAEV_18011981mg; XSha2
Rabbit
Protein A
RUO
16490, 25468
-20°C, Avoid Freeze/Thaw Cycles