missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KV1.1 (KCNA1) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577577
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20C upon receipt. Reconstitute using 50 uL deionized water.
This gene encodes a voltage-gated delayed potassium channel that is phylogenetically related to the Drosophila Shaker channel. The encoded protein has six putative transmembrane segments, and the loop between S5 and S6 forms the pore and contains the conserved selectivity filter motif. The functional channel is a homotetramer. The N-terminus of the channel is associated with beta subunits that can modify the inactivation properties of the channel as well as affect expression levels. The C-terminus of the channel is complexed to a PDZ domain protein that is responsible for channel targeting. Mutations in this gene have been associated with myokymia with periodic ataxia.
Specifications
| KV1.1 (KCNA1) | |
| Polyclonal | |
| Unconjugated | |
| KCNA1 | |
| Kcna1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 16485, 24520, 3736 | |
| -20°C | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.75 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| P10499, P16388, Q09470 | |
| KCNA1 | |
| GST fusion protein with sequence HRETEGEEQAQLLHV SSPNLASDSDLSRRSSSTISKSEYMEIEEDMNNSIAHYRQANIRTGNCTTADQNCVNKSKLLTDV, corresponding to amino acid residues 416-495 of mouse Kv 1.1 | |
| 50 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction