missing translation for 'onlineSavingsMsg'
Learn More

Ku80/XRCC5 Antibody (XRCC5/7315), Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18392856 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392856 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392856 Supplier Novus Biologicals Supplier No. NBP314017JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ku80/XRCC5 Monoclonal antibody specifically detects Ku80/XRCC5 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ku80/XRCC5
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone XRCC5/7315
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias 80kDa, ATP-dependent DNA helicase 2 subunit 2, ATP-dependent DNA helicase II 80 kDa subunit, CTC85, CTCBF, EC 3.6.4.-, FLJ39089, G22P2, KARP1, KARP-1, KU80, Ku86 autoantigen related protein 1,86 kDa subunit of Ku antigen, Ku86DNA repair protein XRCC5, KUB2, NFIV, Thyroid-lupus autoantigen, TLAA, X-ray repair complementing defective repair in Chinese hamster cells 5(double-strand-break rejoining)CTC box-binding factor 85 kDa subunit, X-ray repair complementing defective repair in Chinese hamster cells 5(double-strand-break rejoining; Ku autoantigen, 80kD), X-ray repair cross-complementing protein 5, X-ray repair, complementing defective, repair in Chinese hamster
Host Species Mouse
Immunogen Recombinant fragment (around aa300-500) of human Ku80/XRCC5 (exact sequence is proprietary) (Uniprot: P13010 )
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7520
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.