missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ku80/XRCC5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£145.00 - £330.00
Specifications
| Antigen | Ku80/XRCC5 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) |
| Applications | Western Blot, Immunoprecipitation, ChIP Assay |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18020684
|
Novus Biologicals
NB100-508SS |
0.025 mL |
£145.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18714983
|
Novus Biologicals
NB100-508 |
£330.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Ku80/XRCC5 Polyclonal specifically detects Ku80/XRCC5 in Human, Mouse, Rat, Hamster samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Ku80/XRCC5 | |
| Western Blot 1:500, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) | |
| Polyclonal | |
| Rabbit | |
| Cancer, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining, Stem Cell Markers | |
| PBS with 0.02% Sodium Azide | |
| 7520 | |
| A synthetic peptide corresponding to residues 419-440 LVYVQLPFMEDLRQYMFSSLKN] of the 80 kDa Ku80 protein. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, Immunoprecipitation, ChIP Assay | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rat, Hamster | |
| 80kDa, ATP-dependent DNA helicase 2 subunit 2, ATP-dependent DNA helicase II 80 kDa subunit, CTC85, CTCBF, EC 3.6.4.-, FLJ39089, G22P2, KARP1, KARP-1, KU80, Ku86 autoantigen related protein 1,86 kDa subunit of Ku antigen, Ku86DNA repair protein XRCC5, KUB2, NFIV, Thyroid-lupus autoantigen, TLAA, X-ray repair complementing defective repair in Chinese hamster cells 5(double-strand-break rejoining)CTC box-binding factor 85 kDa subunit, X-ray repair complementing defective repair in Chinese hamster cells 5(double-strand-break rejoining; Ku autoantigen, 80kD), X-ray repair cross-complementing protein 5, X-ray repair, complementing defective, repair in Chinese hamster | |
| XRCC5 | |
| IgG | |
| Affinity Purified | |
| Reacts with residues 419-440 [LVYVQLPFMEDLRQYMFSSLKN] of the 80 kDa Ku80 protein. Sequence is 100% conserved between human, mouse, rat and hamster. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title