Learn More
Invitrogen™ Ku80 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120960
Description
Positive test controls include: A-549, MCF7. The target is usually found in the following locations: Chromosome, Nucleus, nucleolus. Immunogen sequence: KKKDQVTAQE IFQDNHEDGP TAKKLKTEQG GAHFSVSSLA EGSVTSVGSV NPAENFRVLV KQKKASFEEA SNQLINHIEQ FLDTNETPYF MKSIDCIRAF REEAIKFSEE QRFNNFLKAL QEKVEIKQLN HFWEIVVQDG ITLITKEEAS GSSVTAEEAK KFLAPKDKPS GDTAAVFEEG GDVDDLLDMI.
The protein encoded by this gene is the 80-kilodalton subunit of the Ku heterodimer protein which is also known as ATP-dependant DNA helicase II or DNA repair protein XRCC5. Ku is the DNA-binding component of the DNA-dependent protein kinase, and it functions together with the DNA ligase IV-XRCC4 complex in the repair of DNA double-strand break by non-homologous end joining and the completion of V(D)J recombination events. This gene functionally complements Chinese hamster xrs-6, a mutant defective in DNA double-strand break repair and in ability to undergo V(D)J recombination. A rare microsatellite polymorphism in this gene is associated with cancer in patients of varying radiosensitivity.
Specifications
| Ku80 | |
| Polyclonal | |
| Unconjugated | |
| XRCC5 | |
| 86 kDa subunit of Ku antigen; AI314015; ATP-dependent DNA helicase 2 subunit 2; ATP-dependent DNA helicase II 80 kDa subunit; CTC box-binding factor 85 kDa subunit; CTC85; CTCBF; DNA repair protein XRCC5; FLJ39089; G22p2; HGNC:12833; KARP1; KARP-1; Ku autoantigen; ku autoantigen protein p86 homolog; Ku autoantigen, 80kD); Ku autoantigen, 80kDa; Ku p80; Ku80; Ku-80; Ku86; Ku-86; Ku86 autoantigen related protein 1; KUB2; Kup80; Lupu; lupus; lupus Ku autoantigen protein p86; NFIV; nuclear factor IV; OTTHUMP00000164061; OTTHUMP00000206791; Thyroid-lupus autoantigen; TLAA; X-ray repair complementing defective repair in Chinese hamster cells 5; X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining); X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining; Ku autoantigen, 80kDa); X-ray repair cross complementation (double-strand-break rejoining; X-ray repair cross complementation (double-strand-break rejoining; Ku autoantigen, 80kD); X-ray repair cross complementing 5; X-ray repair cross-complementing protein 5; Xrcc5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22596, 363247, 7520 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P13010, P27641 | |
| XRCC5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 543-732 of human Ku80 (NP_066964.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.