missing translation for 'onlineSavingsMsg'
Learn More

Ku70/XRCC6 Antibody [PE], Novus Biologicals™

Product Code. 30651257 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30651257 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30651257 Supplier Novus Biologicals Supplier No. AF5597P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Ku70/XRCC6 Polyclonal antibody specifically detects Ku70/XRCC6 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ku70/XRCC6
Applications Western Blot
Classification Polyclonal
Conjugate PE
Formulation PBS
Gene Alias 5'-deoxyribose-5-phosphate lyase Ku70, 5'-dRP lyase Ku70, 70 kDa subunit of Ku antigen, ATP-dependent DNA helicase 2 subunit 1, ATP-dependent DNA helicase II 70 kDa subunit, CTC box binding factor 75 kDa subunit, CTC box-binding factor 75 kDa subunit, CTC75, CTCBF, D22S731, EC 3.6.4.-, EC 4.2.99.-, G22P1Ku70, Ku autoantigen p70 subunit, Ku autoantigen, 70kDa, KU70, ML8,70 kDa subunit, Thyroid-lupus autoantigen, thyroid-lupus autoantigen p70, TLAAD22S671, X-ray repair complementing defective repair in Chinese hamster cells 6DNA repair protein XRCC6, X-ray repair cross-complementing protein 6
Host Species Goat
Immunogen E. coli-derived recombinant human Ku70, Asn405-Lys556, Accession # P12956
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining
Primary or Secondary Primary
Gene ID (Entrez) 2547
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.