missing translation for 'onlineSavingsMsg'
Learn More

Ku70/XRCC6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-38567-25ul

 View more versions of this product

Product Code. 18658485

  • £240.00 / 25µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Ku70/XRCC6 Polyclonal specifically detects Ku70/XRCC6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ku70/XRCC6
Polyclonal
Western Blot 0.04 - 0.4 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Knockdown Validated
P12956
XRCC6
This antibody was developed against a recombinant protein corresponding to amino acids: PPYFVALVPQEEELDDQKIQVTPPGFQLVFLPFADDKRKMPFTEKIMATPEQVGKMKAIVEKLRFTYRSDSFENPVLQQHFRNLEALALDLMEPEQAVDL
25 μL
Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining
2547
Human
IgG
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
5'-deoxyribose-5-phosphate lyase Ku70, 5'-dRP lyase Ku70, 70 kDa subunit of Ku antigen, ATP-dependent DNA helicase 2 subunit 1, ATP-dependent DNA helicase II 70 kDa subunit, CTC box binding factor 75 kDa subunit, CTC box-binding factor 75 kDa subunit, CTC75, CTCBF, D22S731, EC 3.6.4.-, EC 4.2.99.-, G22P1Ku70, Ku autoantigen p70 subunit, Ku autoantigen, 70kDa, KU70, ML8,70 kDa subunit, Thyroid-lupus autoantigen, thyroid-lupus autoantigen p70, TLAAD22S671, X-ray repair complementing defective repair in Chinese hamster cells 6DNA repair protein XRCC6, X-ray repair cross-complementing protein 6
Rabbit
Affinity Purified
RUO
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.