missing translation for 'onlineSavingsMsg'
Learn More

KU-60019, MedChemExpress

Product Code. 16497697
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497697 5 mg 5mg
16485736 100 mg 100mg
16424968 10 mg 10mg
16401217 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497697 Supplier Medchem Express Supplier No. HY12061
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress KU-60019 is an improved ATM kinase-specific inhibitor with IC50 of 6.3 nM.

KU-60019 is an improved ATM kinase-specific inhibitor with IC50 of 6.3 nM.

Specifications

Chemical Name or Material KU-60019
CAS 925701-46-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C30H33N3O5S
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (182.59 mM; Need ultrasonic)
SMILES C[C@@H]1O[C@@H](CN(CC(NC2=CC3=C(C=C2)SC4=C(C3)C=CC=C4C5=CC(C=C(O5)N6CCOCC6)=O)=O)C1)C
Molecular Weight (g/mol) 547.67
Formula Weight 547.67
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.