missing translation for 'onlineSavingsMsg'
Learn More

KU 55933, TRC

Product Code. 30396125
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30396125 50 mg -
30389512 5 mg -
30389511 10 mg 10mg
3 options
This item is not returnable. View return policy

Product Code. 30396125

Brand: Toronto Research Chemicals TRCK66043850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 587871-26-9
InChI Formula InChI=1S/C21H17NO3S2/c23-14-12-16(25-20(13-14)22-8-10-24-11-9-22)15-4-3-7-19-21(15)27-18-6-2-1-5-17(18)26-19/h1-7,12-13H,8-11H2
Molecular Formula C21H17NO3S2
Molecular Weight (g/mol) 395.49
SMILES O=C1C=C(OC(=C1)c2cccc3Sc4ccccc4Sc23)N5CCOCC5
Synonym KU 55933, ATM Kinase Inhibitor, 2-Thianthren-1-yl-6-(morpholin-4-yl)-4H-pyran-4-one
Chemical Name or Material 2-(4-Morpholinyl)-6-(1-thianthrenyl)-4H-pyran-4-one

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.