missing translation for 'onlineSavingsMsg'
Learn More

KU-0058948, TRC

Product Code. 30392558
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
100 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392558 10 mg -
30396311 100 mg -
30398095 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392558 Supplier Toronto Research Chemicals Supplier No. TRCK66040010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 763111-49-5
Molecular Formula C21 H21 F N4 O2
Molecular Weight (g/mol) 380.42
Synonym 1(2H)-Phthalazinone, 4-[[4-fluoro-3-[(hexahydro-1H-1,4-diazepin-1-yl)carbonyl]phenyl]methyl]-, 1H-1,4-Diazepine, 1-[5-[(3,4-dihydro-4-oxo-1-phthalazinyl)methyl]-2-fluorobenzoyl]hexahydro- (9CI), 4-[[4-Fluoro-3-[(hexahydro-1H-1,4-diazepin-1-yl)carbonyl]phenyl]methyl]-1(2H)-phthalazinone, KU 0058948, KU 0058958
IUPAC Name 4-[[3-(1,4-diazepane-1-carbonyl)-4-fluorophenyl]methyl]-2H-phthalazin-1-one
SMILES Fc1ccc(CC2=NNC(=O)c3ccccc23)cc1C(=O)N4CCCNCC4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H21FN4O2/c22-18-7-6-14(12-17(18)21(28)26-10-3-8-23-9-11-26)13-19-15-4-1-2-5-16(15)20(27)25-24-19/h1-2,4-7,12,23H,3,8-11,13H2,(H,25,27)
Formula Weight 380.1649
Chemical Name or Material KU-0058948
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.