Learn More
Invitrogen™ KTN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588557
Description
Immunogen sequence: MEFYESAYFI VLIPSIVITV IFLFFWLFMK ETLYDEVLAK QKREQKLIPT KTDKKKAEKK KNKKKEIQNG NLHESDSESV PRDFKLSDAL AVEDDQVAPV; Positive Samples: K-562, SKOV3, Raji, HeLa, HT-1080, Mouse heart, Rat testis; Cellular Location: Endoplasmic reticulum membrane, Single-pass type II membrane protein.
Various cellular organelles and vesicles are transported along the microtubules in the cytoplasm. Likewise, membrane recycling of the endoplasmic reticulum (ER), Golgi assembly at the microtubule organizing center, and alignment of lysosomes along microtubules are all related processes. The transport of organelles requires a special class of microtubule-associated proteins (MAPs). One of these is the molecular motor kinesin an ATPase that moves vesicles unidirectionally toward the plus end of the microtubule. Another such MAP is kinectin, a large integral ER membrane protein. Antibodies directed against kinectin have been shown to inhibit its binding to kinesin.
Specifications
| KTN1 | |
| Polyclonal | |
| Unconjugated | |
| KTN1 | |
| CG1; CG-1 antigen; KIAA0004; Kinectin; kinectin 1; kinectin 1 (kinesin receptor); kinesin receptor; KNT; ktn1; MU-RMS-40.19 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 16709, 361029, 3895 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q61595, Q86UP2 | |
| KTN1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human KTN1 (NP_0010729891). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.