missing translation for 'onlineSavingsMsg'
Learn More

KT185, MedChemExpress

Product Code. 16477597
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477597 5 mg 5mg
16474848 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16477597 Supplier Medchem Express Supplier No. HY114926
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress KT185 is an orally-bioavailable, brain-penetrant and selective ABHD6 inhibitor, with an IC50 0.21 nM in Neuro2A cells.

KT185 is an orally-bioavailable, brain-penetrant and selective ABHD6 inhibitor, with an IC50 0.21 nM in Neuro2A cells.

Specifications

Chemical Name or Material KT185
CAS 1472640-86-0
Color White
Purity Grade Notes Research
Molecular Formula C32H33N5O2
Quantity 5 mg
SMILES O=C(N1N=NC(C2=CC=C(C=C2)C3=CC=CC(C(N4CCCCC4)=O)=C3)=C1)N5C(CCCC5)C6=CC=CC=C6
Molecular Weight (g/mol) 519.64
Formula Weight 519.64
Percent Purity 98.78%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.