missing translation for 'onlineSavingsMsg'
Learn More

KT182, MedChemExpress

Product Code. 16487667
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487667 5 mg 5mg
16484918 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16487667 Supplier Medchem Express Supplier No. HY120177
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress KT182 is a potent and selective inhibitor of α/β-hydrolase domain containing 6 (ABHD6), with an IC50 of 0.24 nM in Neuro2A cells.

KT182 is a potent and selective inhibitor of α/β-hydrolase domain containing 6 (ABHD6), with an IC50 of 0.24 nM in Neuro2A cells.

Specifications

Chemical Name or Material KT182
CAS 1402612-62-7
Color White
Purity Grade Notes Research
Molecular Formula C27H26N4O2
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (228.04 mM; Need ultrasonic) ∣Ethanol : 11 mg/mL (25.08 mM; Need ultrasonic and warming)
SMILES O=C(N1N=NC(C2=CC=C(C3=CC=CC(CO)=C3)C=C2)=C1)N4C(C5=CC=CC=C5)CCCC4
Molecular Weight (g/mol) 438.52
Formula Weight 438.52
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.