missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KSR1 Antibody (JE57-75), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32512
This item is not returnable.
View return policy
Description
KSR1 Monoclonal antibody specifically detects KSR1 in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| KSR1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 | |
| kinase suppressor of ras, kinase suppressor of ras 1, KSRRSU2 | |
| Synthetic peptide within Human KSR1 aa 874-923/ 923. (Uniprot: Q8IVT5) | |
| 100 μL | |
| Neuroscience, Phospho Specific, Protein Kinase, Signal Transduction | |
| 8844 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| JE57-75 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction