missing translation for 'onlineSavingsMsg'
Learn More

Ansell KSR™ 22-515 Vinyl Coated Gloves with Interlock Knit Liner

Product Code. 12690772 Shop All Ansell Products
Click to view available options
Size:
7
7.5
8
9
10
Unit Size:
12 pairs
This item is not returnable. View return policy

Product Code. 12690772

Brand: Ansell 22515100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These light, comfortable gloves are ideal for periods of extended wear during light-duty work. Vinyl coating enhances grip when working with small, difficult objects.

  • Slip on cuff for easy donning and doffing
  • Ideal for long wear during light-duty applications
  • Enhanced grip for handling small, difficult objects
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Coating Material PVC
Color Tan
Cut Resistance Rating ANSI A1
Exterior Finish No Texture
Hand Specific
Latex Free Yes
Material Cotton
Product Type Dip-coated Gloves
Size 10
EN388 Standard 2121A
For Use With (Application) Inspection, Assembly, and Metal Fabrication
Glove Cuff Style Open
Liner Material Cotton
Series 22-515
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.