missing translation for 'onlineSavingsMsg'
Learn More

KSHV ORF62 Sheep anti-Virus, Polyclonal, Novus Biologicals™

Product Code. 18331962 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
25 μg
250 μg
Unit Size:
250µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18331962 250 μg 250µg
18345673 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18331962 Supplier Novus Biologicals Supplier No. NBP311885250UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

KSHV ORF62 Polyclonal antibody specifically detects KSHV ORF62 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen KSHV ORF62
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dot Blot
Formulation PBS
Gene Alias Kaposi's Sarcoma Virus, Kaposi's sarcoma-associated herpesvirus, ORF62
Host Species Sheep
Immunogen A fusion protein that contains at its amino terminus the maltose binding protein and at its carboxy terminus the protein encoded by the open reading frame 62 (ORF62) (Uniprot: Q2HR66)
Purification Method Ammonium sulfate precipitation
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4961461
Target Species Virus
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.