missing translation for 'onlineSavingsMsg'
Learn More

KSHV ORF62 Antibody (5B7B6), Novus Biologicals™

Mouse Monoclonal Antibody

£164.00 - £392.00

Specifications

Antigen KSHV ORF62
Clone 5B7B6
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18670776
View Documents
Novus Biologicals
NBP2-37684-0.025ml
0.025 mL
£164.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18196618
View Documents
Novus Biologicals
NBP2-37684
0.1 mL
£392.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

KSHV ORF62 Monoclonal specifically detects KSHV ORF62 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

KSHV ORF62
Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200 - 1:1000
Monoclonal
Ascites
RUO
Ascites with 0.03% Sodium Azide
4961461
IgM
Unpurified
5B7B6
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Virus
Kaposi's Sarcoma Virus, Kaposi's sarcoma-associated herpesvirus
Purified recombinant fragment of human KSHV ORF62 expressed in E. Coli.
Primary
KSHV
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.