missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KSHV ORF45 Antibody (2D4A5), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£187.00 - £446.00
Specifications
| Antigen | KSHV ORF45 |
|---|---|
| Clone | 2D4A5 |
| Dilution | Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18195628
|
Novus Biologicals
NBP2-37685 |
0.1 mL |
£446.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690166
|
Novus Biologicals
NBP2-37685-0.025ml |
0.025 mL |
£187.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KSHV ORF45 Monoclonal specifically detects KSHV ORF45 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| KSHV ORF45 | |
| Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Ascites with 0.03% Sodium Azide | |
| Purified recombinant fragment of KSHV ORF45 expressed in E. Coli. | |
| Primary | |
| KSHV |
| 2D4A5 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Virus | |
| 4961474 | |
| IgG1 | |
| Unpurified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title