missing translation for 'onlineSavingsMsg'
Learn More

Kryptofix™ 222, 98%

Product Code. 10561511 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10561511

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23978-09-8 | C18H36N2O6 | 376.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23978-09-8
Molecular Formula C18H36N2O6
Molecular Weight (g/mol) 376.49
MDL Number MFCD00005111
InChI Key AUFVJZSDSXXFOI-UHFFFAOYSA-N
Synonym kryptofix 222, 4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo 8.8.8 hexacosane, cryptating agent 222, cryptand 222, cryptofix 222, kriptofix 222, cryptate 222, ligand 222, kryptand 222, cryptand c 222
PubChem CID 72801
IUPAC Name 4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane
SMILES C1COCCN2CCOCCOCCN(CCO1)CCOCCOCC2

Specifications

Melting Point 68.0°C to 72.0°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Merck Index 15,2596
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone,diethylether and,chloroform
Formula Weight 376.48
Percent Purity 98%
Chemical Name or Material Kryptofix* 222
compliance-icons
Product Identifier
  • Kryptofix 222
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Kryptofix 222

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.