missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRTHA3B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31709
This item is not returnable.
View return policy
Description
KRTHA3B Polyclonal specifically detects KRTHA3B in Human samples. It is validated for Western Blot.
Specifications
| KRTHA3B | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| KRT33B | |
| Recombinant protein encompassing a sequence within the center region of human KRTHA3B. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 3884 | |
| Human | |
| IgG |
| Western Blot | |
| 0.44mg/mL | |
| Western Blot 1:500-1:3000 | |
| Ha-3II, HA3II, Hair keratin, type I Ha3-II, hard keratin, type I, 3II, hHa3-II, HKA3B, K33B, keratin 33B, keratin, hair, acidic, 3B, keratin, type I cuticular Ha3-II, keratin-33B, KRTHA3A, KRTHA3B, type I hair keratin 3B | |
| Rabbit | |
| 46 kDa | |
| 0.1 mL | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction