missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: HTVWLLEPTC CDNCPPPCHI PQPCV Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
This protein is a member of the keratin-associated protein (KAP) family. The KAP proteins form a matrix of keratin intermediate filaments which contribute to the structure of hair fibers. KAP family members appear to have unique, family-specific amino- and carboxyl-terminal regions and are subdivided into three multi-gene families according to amino acid composition: the high sulfur, the ultrahigh sulfur, and the high tyrosine/glycine KAPs. This protein is a member of the high sulfur KAP family and the gene is localized to a cluster of KAPs at 17q12-q21.
Specifications
Specifications
| Antigen | KRTAP3-3 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | KRTAP3-3 |
| Gene Accession No. | Q9BYR6 |
| Gene Alias | 2310020D06Rik; high sulfur keratin-associated protein 3.3; KAP3.3; keratin associated protein 3-2; keratin associated protein 3-3; keratin-associated protein 3.3; keratin-associated protein 3-2; Keratin-associated protein 3-3; KRTAP3.3; Krtap3-2; krtap3-3; Unknown (protein for MGC:139935) |
| Gene Symbols | KRTAP3-3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?