missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KRTAP24-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564971
This item is not returnable.
View return policy
Description
Immunogen sequence: GSMSTTGYPGV CSTTSYRTHC YIPVTSSVTL SSSDLSPTFG HCLPSSYQGN LWLLDYCQES YGEAPTCKSP S Highest antigen sequence identity to the following orthologs - mouse 67%, rat 67%.
In the hair cortex, hair keratin intermediate filaments are embedded in an interfilamentous matrix, consisting of hair keratin-associated proteins (KRTAP), which are essential for the formation of a rigid and resistant hair shaft through their extensive disulfide bond cross-linking with abundant cysteine residues of hair keratins. The matrix proteins include the high-sulfur and high-glycine-tyrosine keratins.
Specifications
| KRTAP24-1 | |
| Polyclonal | |
| Unconjugated | |
| KRTAP24-1 | |
| KAP24.1; keratin associated protein 24-1; keratin-associated protein 24-1; KRTAP24-1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 643803 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q3LI83 | |
| KRTAP24-1 | |
| Recombinant Human KRTAP24-1. Recombinant protein control fragment (Product #RP-105744). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction