missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRTAP13-1 Antibody (2A1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00140258-M01
This item is not returnable.
View return policy
Description
KRTAP13-1 Monoclonal antibody specifically detects KRTAP13-1 in Human samples. It is validated for Western Blot, ELISA
Specifications
| KRTAP13-1 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| high sulfur keratin associated protein 13.1, KAP13.1High sulfur keratin-associated protein 13.1, keratin associated protein 13-1, keratin-associated protein 13-1, KRTAP13.1 | |
| KRTAP13-1 (NP_853630.1, 2 a.a. ∽ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. WHANSESSQCNSAELTSPINMSYNCCSGNFSSRSCGGYLHYPASSCGFSYPSNQVYSTDLCSPSTCQLGSSLYRGCQQTCWEPTSCQTSYVESSPCQTS | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 2A1 | |
| Western Blot 1:500, ELISA | |
| NP_853630 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 140258 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction