missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KRT9 Recombinant Rabbit Monoclonal Antibody (9A5H3)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA538192
This item is not returnable.
View return policy
Description
Positive Samples: SH-SY5Y Immunogen sequence: DADINGLRQV LDNLTMEKSD LEMQYETLQE ELMALKKNHK EEMSQLTGQN SGDVNVEINV APGKDLTKTL NDMRQEYEQL IAKNRKDIEN QYETQITQIE HEVSSSGQEV QSSAKEVTQL RHGVQELEIE LQSQLSKKAA LEKSLEDTKN RYCGQLQMI.
Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between 40 kDa and 68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium.
Specifications
| KRT9 | |
| Recombinant Monoclonal | |
| 0.38 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35527, Q6RHW0, Q8CIS9 | |
| KRT9 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 9A5H3 | |
| Unconjugated | |
| KRT9 | |
| CK-9; cytokeratin 9; cytokeratin-9; EPPK; K9; keratin 9; keratin 9 (epidermolytic palmoplantar keratoderma); keratin 9, type I; keratin complex 1, acidic, gene 9; keratin, type I cytoskeletal 9; Keratin-9; Krt1-9; Krt9; spermatid perinuclear ring manchette protein K9; type I cytoskeletal 9 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 107656, 266717, 3857 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction