missing translation for 'onlineSavingsMsg'
Learn More

KRT9 Rabbit anti-Human, Mouse, Rat, Clone: 9A5H3, Novus Biologicals™

Product Code. 18340702 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18340702 100 μg 100µL
18335842 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18340702 Supplier Bio-Techne Supplier No. NBP316841100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

KRT9 Monoclonal antibody specifically detects KRT9 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT9
Applications Western Blot
Classification Monoclonal
Clone 9A5H3
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias cytokeratin-9, EPPK, keratin 9, keratin, type I cytoskeletal 9
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 254-412 of human KRT9 (P35527).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3857
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.