missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KRT9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103449
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Cytokeratin 9.
Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between 40 kDa and 68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium.
Specifications
| KRT9 | |
| Polyclonal | |
| Unconjugated | |
| KRT9 | |
| CK-9; cytokeratin 9; cytokeratin-9; EPPK; K9; keratin 9; keratin 9 (epidermolytic palmoplantar keratoderma); keratin 9, type I; keratin complex 1, acidic, gene 9; keratin, type I cytoskeletal 9; Keratin-9; Krt1-9; Krt9; spermatid perinuclear ring manchette protein K9; type I cytoskeletal 9 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 107656, 266717, 3857 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P35527, Q6RHW0, Q8CIS9 | |
| KRT9 | |
| A synthesized peptide derived from human KRT9(Accession P35527), corresponding to amino acid residues R374-T424. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction