Learn More
Invitrogen™ KRT9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587647
Description
Immunogen sequence: TANEKSTMQE LNSRLASYLD KVQALEEANN DLENKIQDWY DKKGPAAIQK NYSPYYNTID DLKDQIVDLT VGNNKTLLDI DNTRMTLDDF RIKFEMEQNL RQGVDADING LRQVLDNLTM EKSDLEMQYE TLQEELMALK KNHKEEMSQL TGQNSGDVNV EINVAPGKDL TKTLNDMRQE YEQLIAKNRK DIENQYETQI TQIEHEVSSS GQEVQSSAKE VTQLRHGVQE LEIELQSQLS KKAALEKSLE DTKNRYCGQL QMIQEQISNL EAQITDVRQE IECQNQEYSL LLSIKMRLEK E; Positive Samples: HeLa, LO2, U-251MG, Mouse liver, Mouse testis, Rat testis.
Cytokeratins are a subfamily of intermediate filament proteins and are characterized by a remarkable biochemical diversity, represented in human epithelial tissues by at least 20 different polypeptides. They range in molecular weight between 40 kDa and 68 kDa and isoelectric pH between 4.9 - 7.8. The individual human cytokeratins are numbered 1 to 20. The various epithelia in the human body usually express cytokeratins which are not only characteristic of the type of epithelium, but also related to the degree of maturation or differentiation within an epithelium.
Specifications
| KRT9 | |
| Polyclonal | |
| Unconjugated | |
| KRT9 | |
| CK-9; cytokeratin 9; cytokeratin-9; EPPK; K9; keratin 9; keratin 9 (epidermolytic palmoplantar keratoderma); keratin 9, type I; keratin complex 1, acidic, gene 9; keratin, type I cytoskeletal 9; Keratin-9; Krt1-9; Krt9; spermatid perinuclear ring manchette protein K9; type I cytoskeletal 9 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 107656, 3857 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.42 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35527, Q6RHW0 | |
| KRT9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 150-450 of human Cytokeratin 9 (KRT9) (KRT9) (NP_0002172). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.