missing translation for 'onlineSavingsMsg'
Learn More

KRT81 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18304555 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18304555 20 μL 20µL
18378636 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18304555 Supplier Novus Biologicals Supplier No. NBP30488020UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

KRT81 Polyclonal specifically detects KRT81 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KRT81
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias ghHb1, ghHkb1, Hair keratin K2.9, hard keratin, type II, 1, Hb-1, hHAKB2-1, K81, keratin 81, Keratin, hair, basic, 1MLN 137, keratin, type II cuticular Hb1, Keratin-81, KRTHB1HB1, Metastatic lymph node 137 gene protein, MLN137, Type II hair keratin Hb1, Type-II keratin Kb21
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 240-390 of human KRT81 (NP_002272.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3887
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.