missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT81 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11342-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the keratin gene family. As a type II hair keratin, it is a basic protein which heterodimerizes with type I keratins to form hair and nails. The type II hair keratins are clustered in a region of chromosome 12q13 and are grouped into two distinct subfamilies based on structure similarity. One subfamily, consisting of KRTHB1, KRTHB3, and KRTHB6, is highly related. The other less-related subfamily includes KRTHB2, KRTHB4, and KRTHB5. All hair keratins are expressed in the hair follicle; this hair keratin, as well as KRTHB3 and KRTHB6, is found primarily in the hair cortex. Mutations in this gene and KRTHB6 have been observed in patients with a rare domit hair disease, monilethrix.Specifications
| KRT81 | |
| Polyclonal | |
| Unconjugated | |
| KRT81 | |
| ghHb1, ghHkb1, Hair keratin K2.9, Hb 1, HB1, hHAKB2 1, K81, keratin 81, Keratin, hair, basic, 1, Keratin, type II cuticular Hb1, KRT81, KRTHB1, MLN 137, MLN137, Type II hair keratin Hb1, Type II keratin Kb21 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3887, 407761, 64818 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14533, Q9ERE2 | |
| KRT81 | |
| KRT81 Fusion Protein Ag1890 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction