missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT80 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16835-1-AP-20UL
This item is not returnable.
View return policy
Description
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene′s expression profile shows that it encodes a type II epithelial keratin, although structurally the encoded protein is more like a type II hair keratin. This protein is involved in cell differentiation, localizing near desmosomal plaques in earlier stages of differentiation but then dispersing throughout the cytoplasm in terminally differentiating cells. The type II keratins are clustered in a region of chromosome 12q13. Two transcript variants encoding two different fully functional isoforms have been found for this gene.Specifications
| KRT80 | |
| Polyclonal | |
| Unconjugated | |
| KRT80 | |
| CK 80, Cytokeratin 80, K80, KB20, keratin 80, KRT80, Type II keratin Kb20 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 144501, 315318, 74127 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q0VBK2, Q6IMF1, Q6KB66 | |
| Krt80 | |
| KRT80 Fusion Protein Ag10498 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction