missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT8/KRT18 Mouse anti-Human, Clone: C-51, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB14663.100ug
This item is not returnable.
View return policy
Description
Mouse monoclonal antibody raised against native human KRT8 and KRT18.
- High specificity
- High reactivity
Specifications
| KRT8/KRT18 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by | |
| P05787; P05783 | |
| KRT8 | |
| Cytoskeleton preparation from HeLa cells. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| C-51 | |
| Mouse monoclonal antibody raised against native human KRT8 and KRT18. | |
| In PBS (0.05% BSA, 0.05% sodium azide). | |
| CARD2/CK8/CYK8/K2C8/K8/KO | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 3856/3875 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction