missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KRT75 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114142
This item is not returnable.
View return policy
Description
Immunogen sequence: SFTTSGGHSL GAGLGGSGFS ATSNRGLGGS GSSVKFVSTT SSSQKSYTH For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Dog: 79%; Human: 100%; Rabbit: 86%.
This gene is a member of the type II keratin family clustered on the long arm of chromosome 12. Type I and type II keratins heteropolymerize to form intermediate-sized filaments in the cytoplasm of epithelial cells.
Specifications
| KRT75 | |
| Polyclonal | |
| Unconjugated | |
| KRT75 | |
| 4732468K03Rik; AA589387; CK-75; cytokeratin KRT2-6HF; cytokeratin type II; cytokeratin-75; hK6hf; K6HF; K75; KB18; keratin 75; keratin 75, type II; keratin, type II cytoskeletal 75; Keratin-6 hair follicle; keratin-75; keratinocyte associated protein 1; Krt2-6hf; KRT75; Krtcap1; mK6hf; PFB; type II keratin Kb18; type II keratin-18; type II keratin-K6hf; Type-II keratin Kb18 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 9119 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O95678 | |
| KRT75 | |
| Synthetic peptide directed towards the middle region of human KRT75. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction