missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KRT73 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA548147
This item is not returnable.
View return policy
Description
KRT73 Polyclonal Antibody for Western Blot, IHC (P)
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene encodes a protein that is expressed in the inner root sheath of hair follicles. The type II keratins are clustered in a region of chromosome 12q13.
Specifications
| KRT73 | |
| Polyclonal | |
| Unconjugated | |
| KRT73 | |
| BC067067; CK-73; cytokeratin-73; IRT6IRS3; K6IRS3; K73; Kb36; keratin 6 irs3; keratin 73; keratin 73, type II; keratin, type II cytoskeletal 73; Keratin-73; KRT6IRS3; Krt73; Type II inner root sheath-specific keratin-K6irs3; type II keratin Kb36; type-II keratin Kb36 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 223915, 319101 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q6NXH9, Q86Y46 | |
| KRT73 | |
| KLH conjugated synthetic peptide between 493-522 amino acids from the C-terminal region of human KRT73. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction