missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KRT72 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584694
This item is not returnable.
View return policy
Description
Immunogen sequence: LSGGIGSSSA SFRARVKGSA SFGSKSLSCL GGSRSLALS Highest antigen sequence indentity to the following orthologs: Mouse - 62%, Rat - 62%.
Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells. The type II keratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This gene encodes a type II keratin that is specifically expressed in the inner root sheath of hair follicles. The type II keratins are clustered in a region of chromosome 12q12-q13. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| KRT72 | |
| Polyclonal | |
| Unconjugated | |
| KRT72 | |
| CK-72; cytokeratin-72; K6irs; K6IRS2; K72; KB35; keratin 6, inner root sheath, 2; keratin 72; keratin 72, type II; keratin protein K6irs; keratin, type II cytoskeletal 72; Keratin-72; KRT6; KRT6IRS2; KRT72; Type II inner root sheath-specific keratin-K6irs2; type-II keratin Kb35 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 140807 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14CN4 | |
| KRT72 | |
| Recombinant protein corresponding to Human KRT72. Recombinant protein control fragment (Product #RP-107437). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction