missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KRT71 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583965
This item is not returnable.
View return policy
Description
Immunogen sequence: EFPSPVSISI ISSTSGGSGY GFRPSMVSGG YVANSSNCIS GVCSVRGGEG RSRGSANDYK DTLGKGSSLS APSKKTSR Highest antigen sequence indentity to the following orthologs: Mouse - 83%, Rat - 82%.
K6IRS1 belongs to a family of type II keratins that are specifically expressed in the inner root sheath of hair follicles.
Specifications
| KRT71 | |
| Polyclonal | |
| Unconjugated | |
| KRT71 | |
| CK-71; cytokeratin-71; hK6irs; hK6irs1; HYPT13; K6IRS1; K71; KB34; Keratin 6 irs; keratin 71; keratin 71, type II; keratin, type II cytoskeletal 71; Keratin-71; KRT6IRS; KRT6IRS1; KRT71; Type II inner root sheath-specific keratin-K6irs1; type-II keratin Kb34 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 112802 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q3SY84 | |
| KRT71 | |
| Recombinant protein corresponding to Human KRT71. Recombinant protein control fragment (Product #RP-97765). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction