missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT40 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25398-1-AP-20UL
This item is not returnable.
View return policy
Description
Cytokeratins, a group comprising at least 29 different proteins, are characteristic of epithelial and trichocytic cells. Cytokeratin 5, 6 and 8 are members of the type II neutral-to-basic subfamily. Monoclonal antibodies to cytokeratins are specific markers of epithelial cell differentiation and have been widely used as tools in tumor identification and classification. PCK-26 facilitates the typing of normal, metaplastic and neoplastic cells and it may aid in the discrimination of carcinomas and non-epithelial tumors such as sarcomas, lymphomas and neural tumors. It is also useful in detecting micro-metastases in lymph nodes and other tissues and for determining the origin of poorly differentiated tumors.Specifications
| KRT40 | |
| Polyclonal | |
| Unconjugated | |
| KRT40 | |
| CK 40, Cytokeratin 40, K40, KA36, keratin 40, KRT40, Type I hair keratin Ka36 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 125115 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.37 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q6A162 | |
| KRT40 | |
| KRT40 Fusion Protein Ag21889 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction