missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT36 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14309-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the keratin gene family. This type I hair keratin is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.Specifications
| KRT36 | |
| Polyclonal | |
| Unconjugated | |
| KRT36 | |
| HA6, Hair keratin, type I Ha6, hHa6, HKA6, K36, keratin 36, Keratin, type I cuticular Ha6, KRT36, KRTHA6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16673, 287698, 8689 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B1AQ75, O76013 | |
| Krt36 | |
| KRT36 Fusion Protein Ag5483 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction