missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KRT33A Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | KRT33A |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
KRT33A Polyclonal specifically detects KRT33A in Human samples. It is validated for Western Blot.Specifications
| KRT33A | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| KRTHA3A | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human KRT33A. Peptide sequence NQVLNETRSQYEALVETNRREVEQWFATQTEELNKQVVSSSEQLQSYQAE | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 3883 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit