missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cytokeratin 31/33A/33B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100847
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total KRT31/33A/33B.
The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
Specifications
| Cytokeratin 31/33A/33B | |
| Polyclonal | |
| Unconjugated | |
| KRT33A | |
| HA1; Ha-1; HA3I; Ha-3I; HA3II; Ha-3II; hair keratin; hair keratin, type I Ha1; Hair keratin, type I Ha3-I; hair keratin, type I Ha3-II; hard keratin, type I, 1; hard keratin, type I, 3II; hard keratin, type I,3I; hHa1; HHA3-I; HHA3-II; HKA1; HKA3A; HKA3B; K31; K33A; K33B; keratin 31; keratin 31, type I; keratin 33A; keratin 33A, type I; keratin 33B; keratin 33B, type I; keratin, hair, acidic, 3A; keratin, hair, acidic, 3B; keratin, hair, acidic,1; keratin, type I cuticular Ha1; Keratin, type I cuticular Ha3-I; keratin, type I cuticular Ha3-II; Keratin-31; Keratin-33A; Keratin-33B; Krt1-3; KRT31; KRT33A; KRT33B; KRTHA1; KRTHA3A; KRTHA3B; type I hair keratin 3B | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16660, 16671, 303527, 3881, 3883, 3884, 450227, 450228, 71888 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O76009, Q14525, Q15323, Q61765, Q61897, Q8K0Y2 | |
| KRT31, KRT33A, KRT33B | |
| A synthesized peptide derived from human KRT33A(Accession O76009), corresponding to amino acid residues E304-N354. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction