missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Krt33a Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571519
This item is not returnable.
View return policy
Description
Krt33a Polyclonal Antibody for Western Blot
The protein encoded by this gene is a member of the keratin gene family. It is one of the type I hair keratin genes which are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. There are two isoforms of this protein, encoded by two separate genes, KRTHA3A and KRTHA3B. [provided by RefSeq].
Specifications
| Krt33a | |
| Polyclonal | |
| Unconjugated | |
| Krt33a | |
| 2310015J09Rik; HA3I; Ha-3I; hair keratin; Hair keratin, type I Ha3-I; hard keratin, type I,3I; HHA3-I; HKA3A; K33A; Ka27; keratin 33A; keratin 33A, type I; keratin, hair, acidic, 3A; keratin, type I cuticular Ha3-I; Keratin-33A; Krt1-3; krt33a; KRTHA3A; ortholog of keratin, hair, acidic, 3A KRTHA3A; RGD1306249; type I hair keratin KA27 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 3883, 71888 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.76 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O76009, Q8K0Y2 | |
| Krt33a | |
| KLH conjugated synthetic peptide between 56-85 amino acids from the N-terminal region of human KT33A. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction